Learn More
Medchemexpress LLC Bis-PEG25-NHS ester | 2221948-96-3 | 99.8% | 1413.55 | C62H112N2O33 | 100 MG
SDP
Catalog No. 5000202737
Encompass_Preferred
Bis-PEG25-NHS ester is a bifunctional PEG-based linker bearing two N-hydroxysuccinimide (NHS) ester groups for efficient amine conjugation. It is used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a spacer in PROTAC construction; the PEG25 spacer improves aqueous solubility. Intended for research use only.
  • Contains two NHS ester groups for amine coupling.
  • PEG25 spacer enhances solubility in aqueous media.
  • Suitable for ADC linker and PROTAC synthesis.
  • High purity (99.75%) for reproducible conjugation.
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
Catalog No. 50-002-02737 Supplier Medchemexpress LLC Supplier No. HY130828100MG
Bis-PEG25-NHS ester is a bifunctional PEG-based linker bearing two N-hydroxysuccinimide (NHS) ester groups for efficient amine conjugation. It is used as a cleavable linker in antibody-drug conjugate (ADC) synthesis and as a spacer in PROTAC construction; the PEG25 spacer improves aqueous solubility. Intended for research use only.
  • Contains two NHS ester groups for amine coupling.
  • PEG25 spacer enhances solubility in aqueous media.
  • Suitable for ADC linker and PROTAC synthesis.
  • High purity (99.75%) for reproducible conjugation.
  • Store sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).