Learn More
Medchemexpress LLC Bis-PEG25-NHS ester | 2221948-96-3 | MFCD21363290 | 99.8% | 1413.55 | C62H112N2O33 | 25 MG
SDP
Catalog No. 5000203388
Encompass_Preferred
Bis-PEG25-NHS ester is a PEG/alkyl/ether-based homobifunctional linker containing two N-hydroxysuccinimide (NHS) ester groups, used for conjugation in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs). It is provided as a hydrophilic solid for use in organic solvents and should be stored sealed at -20°C.
  • Contains two amine-reactive NHS ester groups for conjugation.
  • PEG spacer increases hydrophilicity and solubility in aqueous media.
  • Suitable as a cleavable linker for ADC and PROTAC synthesis.
  • Provided as a solid with recommended cold, sealed storage to preserve stability.
Catalog No. 50-002-03388 Supplier Medchemexpress LLC Supplier No. HY13082825MG
Bis-PEG25-NHS ester is a PEG/alkyl/ether-based homobifunctional linker containing two N-hydroxysuccinimide (NHS) ester groups, used for conjugation in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs). It is provided as a hydrophilic solid for use in organic solvents and should be stored sealed at -20°C.
  • Contains two amine-reactive NHS ester groups for conjugation.
  • PEG spacer increases hydrophilicity and solubility in aqueous media.
  • Suitable as a cleavable linker for ADC and PROTAC synthesis.
  • Provided as a solid with recommended cold, sealed storage to preserve stability.