Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG25-NHS ester | 2221948-96-3 | MFCD21363290 | 99.8% | 1413.55 | C62H112N2O33 | 50 MG
SDP

Catalog No. 5000203389
Encompass_Preferred

Bis-PEG25-NHS ester is a bifunctional PEG-based N-hydroxysuccinimide (NHS) ester linker used for bioconjugation, PROTAC linker assembly, and as a cleavable ADC linker. It provides a hydrophilic PEG25 spacer with two NHS reactive groups for amine coupling and conjugation chemistries.

  • Bifunctional PEG25 linker with two NHS ester reactive groups.
  • Purity: 99.75%.
  • Molecular weight: 1413.55.
  • Formula: C62H112N2O33.
  • CAS number: 2221948-96-3.
  • Suitable for PROTAC, ADC, and general bioconjugation applications.
  • Storage: store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month; dry storage at -20°C.

Catalog No. 50-002-03389 Supplier Medchemexpress LLC Supplier No. HY13082850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG25-NHS ester is a bifunctional PEG-based N-hydroxysuccinimide (NHS) ester linker used for bioconjugation, PROTAC linker assembly, and as a cleavable ADC linker. It provides a hydrophilic PEG25 spacer with two NHS reactive groups for amine coupling and conjugation chemistries.

  • Bifunctional PEG25 linker with two NHS ester reactive groups.
  • Purity: 99.75%.
  • Molecular weight: 1413.55.
  • Formula: C62H112N2O33.
  • CAS number: 2221948-96-3.
  • Suitable for PROTAC, ADC, and general bioconjugation applications.
  • Storage: store sealed, away from moisture; in solvent: -80°C up to 6 months, -20°C up to 1 month; dry storage at -20°C.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.