Learn More
Selleck Chemical LLC Bis-PEG3-acid S1350-100mg
SDP
Catalog No. 503110631
Encompass_Preferred
Bis-PEG3-acid is a PEG-based PROTAC linker can be used in the synthesis of PROTACs
Catalog No. 50-311-0631 Supplier Selleck Chemical LLC Supplier No. S1350100MG
Bis-PEG3-acid is a PEG-based PROTAC linker can be used in the synthesis of PROTACs