Learn More
Medchemexpress LLC Bis-PEG3-NHS ester | 1314378-16-9 | 97.0% | 444.39 g/mol | C18H24N2O11 | 500 MG
SDP
Catalog No. 5000203848
Encompass_Preferred
Bis-PEG3-NHS ester is a non-cleavable PEG-based bifunctional linker containing two N-hydroxysuccinimide (NHS) ester reactive groups used for bioconjugation and antibody-drug conjugate (ADC) construction. The three-unit PEG spacer increases hydrophilicity and solubility, while the NHS esters enable efficient coupling to primary amines on proteins and peptides.
  • Non-cleavable 3-unit PEG linker for stable conjugation.
  • Contains two NHS ester groups for bioconjugation to primary amines.
  • Improves solubility and hydrophilicity of conjugates.
  • Available in multiple package sizes, including 500 mg.
  • Provided with SDS, COA, and product data sheet for quality and safety information.
Catalog No. 50-002-03848 Supplier Medchemexpress LLC Supplier No. HY130087500MG
Bis-PEG3-NHS ester is a non-cleavable PEG-based bifunctional linker containing two N-hydroxysuccinimide (NHS) ester reactive groups used for bioconjugation and antibody-drug conjugate (ADC) construction. The three-unit PEG spacer increases hydrophilicity and solubility, while the NHS esters enable efficient coupling to primary amines on proteins and peptides.
  • Non-cleavable 3-unit PEG linker for stable conjugation.
  • Contains two NHS ester groups for bioconjugation to primary amines.
  • Improves solubility and hydrophilicity of conjugates.
  • Available in multiple package sizes, including 500 mg.
  • Provided with SDS, COA, and product data sheet for quality and safety information.