Learn More
Medchemexpress LLC Bis-PEG30-NHS ester | 1633.81 | 25 MG
SDP
Catalog No. 5000395241
Encompass_Preferred
Bis-PEG30-NHS ester is a non-cleavable 30-unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs).
  • Target: ADC linker
  • Pathway: Antibody-drug conjugate/ADC related
Catalog No. 50-003-95241 Supplier Medchemexpress LLC Supplier No. 25MGHY164724
Bis-PEG30-NHS ester is a non-cleavable 30-unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs).
  • Target: ADC linker
  • Pathway: Antibody-drug conjugate/ADC related