Learn More
Medchemexpress LLC Bis-PEG30-NHS ester | 1633.81 | 5 MG
SDP
Catalog No. 5000391734
Encompass_Preferred
Bis-PEG30-NHS ester is a non-cleavable 30-unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It is designed for various laboratory applications.
  • Non-cleavable 30-unit PEG ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
Catalog No. 50-003-91734 Supplier Medchemexpress LLC Supplier No. HY1647245MG
Bis-PEG30-NHS ester is a non-cleavable 30-unit PEG ADC linker used in the synthesis of antibody-drug conjugates (ADCs). It is designed for various laboratory applications.
  • Non-cleavable 30-unit PEG ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)