Learn More
Medchemexpress LLC Bis-PEG4-NHS ester | 1g
SDP
Catalog No. 5000203011
Encompass_Preferred
Bis- PEG4- NHS ester is a nonclaevable 4-unit PEG linker for antibody-agent-conjugation (ADC)
Catalog No. 50-002-03011 Supplier Medchemexpress LLC Supplier No. HY1300861G
Bis- PEG4- NHS ester is a nonclaevable 4-unit PEG linker for antibody-agent-conjugation (ADC)