Learn More
Medchemexpress LLC Bis-PEG4-PFP ester | 1314378-12-5 | 97.0% | 10 G
SDP
Catalog No. 5000342827
Encompass_Preferred
Bis-PEG4-PFP ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • For research use only
Catalog No. 50-003-42827 Supplier Medchemexpress LLC Supplier No. HY12699810G
Bis-PEG4-PFP ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs consist of two different ligands connected by a linker, one for an E3 ubiquitin ligase and another for a target protein. They exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Exploits the ubiquitin-proteasome system
  • Enables selective degradation of target proteins
  • For research use only