Learn More
Medchemexpress LLC Bis-PEG4-PFP ester | 1314378-12-5 | 97.0% | 500 MG
SDP
Catalog No. 5000338695
Encompass_Preferred
Bis-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs consist of two different ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase.
  • The other ligand targets the protein of interest.
  • PROTACs utilize the intracellular ubiquitin-proteasome system.
  • Selectively degrade target proteins.
Catalog No. 50-003-38695 Supplier Medchemexpress LLC Supplier No. HY126998500MG
Bis-PEG4-PFP ester is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs.
  • PROTACs consist of two different ligands connected by a linker.
  • One ligand targets an E3 ubiquitin ligase.
  • The other ligand targets the protein of interest.
  • PROTACs utilize the intracellular ubiquitin-proteasome system.
  • Selectively degrade target proteins.