Learn More
Medchemexpress LLC Bis-PEG4-sulfonic acid | 1807539-08-7 | 366.41 | 100 MG
SDP
Catalog No. 5000349802
Encompass_Preferred
Bis-PEG4-sulfonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • For research use only
Catalog No. 50-003-49802 Supplier Medchemexpress LLC Supplier No. HY140166100MG
Bis-PEG4-sulfonic acid is a PEG-based PROTAC linker used in the synthesis of PROTACs. PROTACs contain two different ligands connected by a linker; one for an E3 ubiquitin ligase and the other for the target protein. These molecules exploit the intracellular ubiquitin-proteasome system to selectively degrade target proteins. This product is for research use only.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Facilitates selective degradation of target proteins
  • For research use only