Learn More
Medchemexpress LLC Bis-PEG4-sulfonic acid | 1807539-08-7 | 366.41 | 25 MG
SDP
Catalog No. 5000351390
Encompass_Preferred
Bis-PEG4-sulfonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This compound appears as a colorless to light yellow liquid.
  • Peg-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Ligands for E3 ubiquitin ligase and target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins
Catalog No. 50-003-51390 Supplier Medchemexpress LLC Supplier No. 25MGHY140166
Bis-PEG4-sulfonic acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. This compound appears as a colorless to light yellow liquid.
  • Peg-based PROTAC linker
  • Used for the synthesis of PROTACs
  • Contains two different ligands connected by a linker
  • Ligands for E3 ubiquitin ligase and target protein
  • Exploits the intracellular ubiquitin-proteasome system
  • Selectively degrades target proteins