Learn More
Medchemexpress LLC Bis-PEG4-sulfonic acid | 1807539-08-7 | 366.41 | 50 MG
SDP
Catalog No. 5000350509
Encompass_Preferred
Bis-PEG4-sulfonic acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These PROTACs are compounds that facilitate the selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system
  • For research use only
Catalog No. 50-003-50509 Supplier Medchemexpress LLC Supplier No. HY14016650MG
Bis-PEG4-sulfonic acid is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. These PROTACs are compounds that facilitate the selective degradation of target proteins by leveraging the intracellular ubiquitin-proteasome system.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Leverages the ubiquitin-proteasome system
  • For research use only