Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG4-TFP ester | 1446282-42-3 | 98.0% | 590.41 | C24H22F8O8 | 100 MG
SDP

Catalog No. 5000281591
Encompass_Preferred

Bis-PEG4-TFP ester is a PEG-based bifunctional linker with two tetrafluorophenyl (TFP) ester reactive termini separated by a PEG4 spacer. It is intended for conjugation in the synthesis of proteolysis-targeting chimeras (PROTACs) and other bifunctional molecules.

  • Provides two TFP ester reactive termini for efficient conjugation
  • PEG4 spacer improves solubility and molecular flexibility
  • High purity suitable for research synthesis (98.0%)
  • Molecular weight 590.41 and CAS 1446282-42-3 for precise identification
  • Available in multiple pack sizes, including 100 mg
  • Store at -20°C, protect from light, under inert atmosphere
  • Suitable for use in PROTAC assembly and linker optimization

Catalog No. 50-002-81591 Supplier Medchemexpress LLC Supplier No. HYW190728100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG4-TFP ester is a PEG-based bifunctional linker with two tetrafluorophenyl (TFP) ester reactive termini separated by a PEG4 spacer. It is intended for conjugation in the synthesis of proteolysis-targeting chimeras (PROTACs) and other bifunctional molecules.

  • Provides two TFP ester reactive termini for efficient conjugation
  • PEG4 spacer improves solubility and molecular flexibility
  • High purity suitable for research synthesis (98.0%)
  • Molecular weight 590.41 and CAS 1446282-42-3 for precise identification
  • Available in multiple pack sizes, including 100 mg
  • Store at -20°C, protect from light, under inert atmosphere
  • Suitable for use in PROTAC assembly and linker optimization

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.