Learn More
Medchemexpress LLC Bis-PEG4-TFP ester | 1446282-42-3 | 98.0% | 590.41 g/mol | C24H22F8O8 | 50 MG
SDP
Catalog No. 5000281574
Encompass_Preferred
Bis-PEG4-TFP ester is a PEG-based PROTAC linker with two tetrafluorophenyl (TFP) activated ester groups that enable facile conjugation to amine-containing ligands. It is provided as a high-purity research reagent intended for use in synthesis of bifunctional molecules and linker optimization.
  • Contains two TFP-activated ester termini for rapid amide formation.
  • Provides a PEG4 spacer to improve solubility and molecular flexibility.
  • Enables efficient coupling to primary amines under mild conditions.
  • High purity suitable for research and synthetic workflows.
  • Available in small pack sizes convenient for laboratory use.
Catalog No. 50-002-81574 Supplier Medchemexpress LLC Supplier No. HYW19072850MG
Bis-PEG4-TFP ester is a PEG-based PROTAC linker with two tetrafluorophenyl (TFP) activated ester groups that enable facile conjugation to amine-containing ligands. It is provided as a high-purity research reagent intended for use in synthesis of bifunctional molecules and linker optimization.
  • Contains two TFP-activated ester termini for rapid amide formation.
  • Provides a PEG4 spacer to improve solubility and molecular flexibility.
  • Enables efficient coupling to primary amines under mild conditions.
  • High purity suitable for research and synthetic workflows.
  • Available in small pack sizes convenient for laboratory use.