Learn More
Medchemexpress LLC Bis-PEG5-acid 10g | 439114-13-3 | 338.35 g/mol | C14H26O9 | 10 G
SDP
Catalog No. 5000226325
Encompass_Preferred
Bis-PEG5-acid is a polyethylene glycol (PEG) linker with two terminal carboxylic acid groups used to assemble PROTACs and other bifunctional molecules. The hydrophilic spacer increases aqueous solubility and provides flexible separation between ligands for conjugation chemistry.
  • Two terminal carboxylic acid groups for conjugation.
  • Hydrophilic PEG spacer improves solubility in aqueous media.
  • Flexible linker suitable for PROTAC and bifunctional molecule assembly.
  • Available in multiple package sizes including 10 G.
  • Solid material with white to yellow appearance.
Catalog No. 50-002-26325 Supplier Medchemexpress LLC Supplier No. HY11600610G
Bis-PEG5-acid is a polyethylene glycol (PEG) linker with two terminal carboxylic acid groups used to assemble PROTACs and other bifunctional molecules. The hydrophilic spacer increases aqueous solubility and provides flexible separation between ligands for conjugation chemistry.
  • Two terminal carboxylic acid groups for conjugation.
  • Hydrophilic PEG spacer improves solubility in aqueous media.
  • Flexible linker suitable for PROTAC and bifunctional molecule assembly.
  • Available in multiple package sizes including 10 G.
  • Solid material with white to yellow appearance.