Learn More
Medchemexpress LLC Bis-PEG5-acid 1g | 439114-13-3 | 338.35 g/mol | C14H26O9 | 1 G
SDP
Catalog No. 5000307672
Encompass_Preferred
Bis-PEG5-acid is a polyethylene glycol (PEG) linker bearing two terminal carboxylic acid groups, used as a hydrophilic spacer in PROTAC construction and bioconjugation chemistry. It facilitates coupling reactions and improves aqueous solubility for assembled molecules.
  • Two terminal carboxylic acid groups enable standard amide coupling reactions.
  • PEG5 spacer provides hydrophilicity and conformational flexibility.
  • Molecular weight 338.35 g/mol and formula C14H26O9 for precise stoichiometry.
  • Suitable for PROTAC synthesis and general linker applications in medicinal chemistry.
  • Offered in research quantities including 1 g for small-scale synthesis.
Catalog No. 50-003-07672 Supplier Medchemexpress LLC Supplier No. HY1160061G
Bis-PEG5-acid is a polyethylene glycol (PEG) linker bearing two terminal carboxylic acid groups, used as a hydrophilic spacer in PROTAC construction and bioconjugation chemistry. It facilitates coupling reactions and improves aqueous solubility for assembled molecules.
  • Two terminal carboxylic acid groups enable standard amide coupling reactions.
  • PEG5 spacer provides hydrophilicity and conformational flexibility.
  • Molecular weight 338.35 g/mol and formula C14H26O9 for precise stoichiometry.
  • Suitable for PROTAC synthesis and general linker applications in medicinal chemistry.
  • Offered in research quantities including 1 g for small-scale synthesis.