Learn More
Medchemexpress LLC Bis-PEG5-acid 5g | 439114-13-3 | 338.35 g·mol⁻1 | C14H26O9 | 5 G
SDP
Catalog No. 5000228263
Encompass_Preferred
Bis-PEG5-acid is a polyethylene glycol (PEG)-based bifunctional linker with two terminal carboxylic acid groups, used as a hydrophilic spacer in PROTAC synthesis, bioconjugation, and medicinal chemistry to improve solubility and provide flexibility between binding motifs.
  • Provides a flexible PEG5 spacer for conjugation and linker design.
  • Two terminal carboxylic acid groups for straightforward coupling chemistry.
  • High purity (99.9%) suitable for research applications.
  • Available in laboratory pack sizes for small-scale synthesis.
  • Enhances aqueous solubility of hydrophobic ligands and intermediates.
Catalog No. 50-002-28263 Supplier Medchemexpress LLC Supplier No. HY1160065G
Bis-PEG5-acid is a polyethylene glycol (PEG)-based bifunctional linker with two terminal carboxylic acid groups, used as a hydrophilic spacer in PROTAC synthesis, bioconjugation, and medicinal chemistry to improve solubility and provide flexibility between binding motifs.
  • Provides a flexible PEG5 spacer for conjugation and linker design.
  • Two terminal carboxylic acid groups for straightforward coupling chemistry.
  • High purity (99.9%) suitable for research applications.
  • Available in laboratory pack sizes for small-scale synthesis.
  • Enhances aqueous solubility of hydrophobic ligands and intermediates.