Learn More
Medchemexpress LLC Bis-PEG5-NHS ester | 756526-03-1 | 99.8% | 532.50 g/mol | C22H32N2O13 | 25 MG
SDP
Catalog No. 5000204198
Encompass_Preferred
Bis-PEG5-NHS ester is a bifunctional PEG-based cleavable NHS-activated ester used for conjugation to primary amines in the synthesis of antibody-drug conjugates (ADCs), PROTACs, and other bioconjugates. Supplied as a solid for research use, it facilitates PEGylation and formation of cleavable linkers in medicinal chemistry and chemical biology workflows.
  • Cleavable PEG5 NHS ester for selective amine conjugation.
  • Suitable for ADC and PROTAC linker synthesis.
  • High purity, suitable for synthetic applications.
  • Molecular weight 532.50 g/mol, chemical formula C22H32N2O13.
  • Available in small-scale research quantities, e.g. 25 MG.
Catalog No. 50-002-04198 Supplier Medchemexpress LLC Supplier No. HY12688925MG
Bis-PEG5-NHS ester is a bifunctional PEG-based cleavable NHS-activated ester used for conjugation to primary amines in the synthesis of antibody-drug conjugates (ADCs), PROTACs, and other bioconjugates. Supplied as a solid for research use, it facilitates PEGylation and formation of cleavable linkers in medicinal chemistry and chemical biology workflows.
  • Cleavable PEG5 NHS ester for selective amine conjugation.
  • Suitable for ADC and PROTAC linker synthesis.
  • High purity, suitable for synthetic applications.
  • Molecular weight 532.50 g/mol, chemical formula C22H32N2O13.
  • Available in small-scale research quantities, e.g. 25 MG.