Learn More
Medchemexpress LLC Bis-PEG6-NHS ester | 1526718-98-8 | 576.55 g/mol | C24H36N2O14 | 1 G
SDP
Catalog No. 5000202718
Encompass_Preferred
Bis-PEG6-NHS ester is a PEG-based bifunctional linker containing two N-hydroxysuccinimide (NHS) ester reactive groups separated by a six-unit polyethylene glycol spacer. It is used for conjugation to primary amines and as a linker in PROTAC and cleavable ADC synthesis.
  • Provides two NHS ester reactive groups for amine coupling.
  • PEG6 spacer improves solubility and flexibility.
  • Suitable for PROTAC and ADC linker synthesis.
  • Molecular weight 576.55 g/mol and formula C24H36N2O14.
  • Available in multiple pack sizes including 1 g.
Catalog No. 50-002-02718 Supplier Medchemexpress LLC Supplier No. HY1304101G
Bis-PEG6-NHS ester is a PEG-based bifunctional linker containing two N-hydroxysuccinimide (NHS) ester reactive groups separated by a six-unit polyethylene glycol spacer. It is used for conjugation to primary amines and as a linker in PROTAC and cleavable ADC synthesis.
  • Provides two NHS ester reactive groups for amine coupling.
  • PEG6 spacer improves solubility and flexibility.
  • Suitable for PROTAC and ADC linker synthesis.
  • Molecular weight 576.55 g/mol and formula C24H36N2O14.
  • Available in multiple pack sizes including 1 g.