Learn More
Medchemexpress LLC Bis-PEG6-NHS ester | 1526718-98-8 | 98.0% | 576.55 g/mol | C24H36N2O14 | 50 MG
SDP
Catalog No. 5000204125
Encompass_Preferred
Bis-PEG6-NHS ester is a PEG-based bifunctional linker bearing two N-hydroxysuccinimide (NHS) ester termini separated by a six-unit polyethylene glycol spacer. It is used to conjugate amine-containing ligands in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs).
  • High purity (98.0%).
  • Two NHS ester termini for efficient amine coupling.
  • PEG6 spacer provides aqueous solubility and conformational flexibility.
  • Suitable as a cleavable linker in ADC construction and for PROTAC linker design.
  • Store sealed at -20 °C, away from moisture; unstable in solution, prepare fresh as needed.
Catalog No. 50-002-04125 Supplier Medchemexpress LLC Supplier No. HY13041050MG
Bis-PEG6-NHS ester is a PEG-based bifunctional linker bearing two N-hydroxysuccinimide (NHS) ester termini separated by a six-unit polyethylene glycol spacer. It is used to conjugate amine-containing ligands in the synthesis of PROTACs and as a cleavable linker for antibody-drug conjugates (ADCs).
  • High purity (98.0%).
  • Two NHS ester termini for efficient amine coupling.
  • PEG6 spacer provides aqueous solubility and conformational flexibility.
  • Suitable as a cleavable linker in ADC construction and for PROTAC linker design.
  • Store sealed at -20 °C, away from moisture; unstable in solution, prepare fresh as needed.