Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG7-NHS ester | 1334170-02-3 | 98.0% | 620.60 g/mol | C26H40N2O15 | 250 MG
SDP

Catalog No. 5000203552
Encompass_Preferred

Bis-PEG7-NHS ester is a bifunctional PEG-based linker bearing two N-hydroxysuccinimide (NHS) ester groups. It is used as a building block in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs), offering a hydrophilic spacer and amine-reactive termini for conjugation chemistry.

  • Contains two NHS ester functional groups for amine coupling.
  • PEG7 spacer increases solubility in aqueous media.
  • Suitable for ADC and PROTAC linker synthesis.
  • Analytical purity: 98.0%.
  • Molecular weight: 620.60 g/mol; molecular formula: C26H40N2O15.
  • Store at -20°C; stable for extended storage under recommended conditions.

Catalog No. 50-002-03552 Supplier Medchemexpress LLC Supplier No. HY126890250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG7-NHS ester is a bifunctional PEG-based linker bearing two N-hydroxysuccinimide (NHS) ester groups. It is used as a building block in the synthesis of antibody-drug conjugates (ADCs) and proteolysis targeting chimeras (PROTACs), offering a hydrophilic spacer and amine-reactive termini for conjugation chemistry.

  • Contains two NHS ester functional groups for amine coupling.
  • PEG7 spacer increases solubility in aqueous media.
  • Suitable for ADC and PROTAC linker synthesis.
  • Analytical purity: 98.0%.
  • Molecular weight: 620.60 g/mol; molecular formula: C26H40N2O15.
  • Store at -20°C; stable for extended storage under recommended conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.