Learn More
Medchemexpress LLC Bis-PEG9-NHS ester 1g | 1008402-79-6 | 1 G
SDP
Catalog No. 5000307604
Encompass_Preferred
Bis-PEG9-NHS ester is a homobifunctional polyethylene glycol (PEG) linker bearing two N-hydroxysuccinimide (NHS) ester groups. It reacts with primary amines to form stable amide bonds and is used for protein conjugation, PROTAC and antibody-drug conjugate (ADC) linker synthesis. Provided as a high-purity research reagent.
  • Homobifunctional PEG9 spacer with two NHS ester reactive groups.
  • Enables amine-targeted conjugation to proteins and small molecules.
  • PEG spacer increases aqueous solubility and reduces aggregation.
  • Suitable for PROTAC and ADC linker synthesis and general bioconjugation.
  • High purity for consistent research results.
Catalog No. 50-003-07604 Supplier Medchemexpress LLC Supplier No. HY1170091G
Bis-PEG9-NHS ester is a homobifunctional polyethylene glycol (PEG) linker bearing two N-hydroxysuccinimide (NHS) ester groups. It reacts with primary amines to form stable amide bonds and is used for protein conjugation, PROTAC and antibody-drug conjugate (ADC) linker synthesis. Provided as a high-purity research reagent.
  • Homobifunctional PEG9 spacer with two NHS ester reactive groups.
  • Enables amine-targeted conjugation to proteins and small molecules.
  • PEG spacer increases aqueous solubility and reduces aggregation.
  • Suitable for PROTAC and ADC linker synthesis and general bioconjugation.
  • High purity for consistent research results.