Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-PEG9-NHS ester 250mg | 1008402-79-6 | 250 MG
SDP

Catalog No. 5000307608
Encompass_Preferred

Bis-PEG9-NHS ester is a homobifunctional polyethylene glycol (PEG) linker bearing two N-hydroxysuccinimide (NHS) ester reactive groups, used for amine-directed bioconjugation in the synthesis of PROTACs and antibody-drug conjugates. The PEG9 spacer increases hydrophilicity to improve solubility and reduce steric hindrance during conjugation.

  • Contains two NHS ester reactive groups for amine coupling.
  • PEG9 spacer increases hydrophilicity and solubility in aqueous media.
  • High purity (≥98.0%) suitable for research and conjugation chemistry.
  • Molecular weight 708.71 g/mol and chemical formula C30H48N2O17.
  • Viscous, colorless to light yellow appearance for easy handling.
  • Applicable to protein labeling, bioconjugation, and linker synthesis.

Catalog No. 50-003-07608 Supplier Medchemexpress LLC Supplier No. HY117009250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-PEG9-NHS ester is a homobifunctional polyethylene glycol (PEG) linker bearing two N-hydroxysuccinimide (NHS) ester reactive groups, used for amine-directed bioconjugation in the synthesis of PROTACs and antibody-drug conjugates. The PEG9 spacer increases hydrophilicity to improve solubility and reduce steric hindrance during conjugation.

  • Contains two NHS ester reactive groups for amine coupling.
  • PEG9 spacer increases hydrophilicity and solubility in aqueous media.
  • High purity (≥98.0%) suitable for research and conjugation chemistry.
  • Molecular weight 708.71 g/mol and chemical formula C30H48N2O17.
  • Viscous, colorless to light yellow appearance for easy handling.
  • Applicable to protein labeling, bioconjugation, and linker synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.