Learn More
Medchemexpress LLC Bis-PEG9-NHS ester 5g | 1008402-79-6 | 708.71 | C30H48N2O17 | 5 G
SDP
Catalog No. 5000307606
Encompass_Preferred
Bis-PEG9-NHS ester is a homobifunctional PEG-based linker bearing two N-hydroxysuccinimide (NHS) ester reactive groups, used to conjugate primary amines on proteins, peptides, and small molecules for research applications.
  • Homobifunctional PEG9 spacer with two NHS ester reactive groups.
  • Facilitates amine-directed bioconjugation to proteins and peptides.
  • Hydrophilic PEG spacer improves solubility in aqueous media.
  • Suitable for PROTAC and ADC linker chemistry in research.
  • High purity for reliable experimental performance.
Catalog No. 50-003-07606 Supplier Medchemexpress LLC Supplier No. HY1170095G
Bis-PEG9-NHS ester is a homobifunctional PEG-based linker bearing two N-hydroxysuccinimide (NHS) ester reactive groups, used to conjugate primary amines on proteins, peptides, and small molecules for research applications.
  • Homobifunctional PEG9 spacer with two NHS ester reactive groups.
  • Facilitates amine-directed bioconjugation to proteins and peptides.
  • Hydrophilic PEG spacer improves solubility in aqueous media.
  • Suitable for PROTAC and ADC linker chemistry in research.
  • High purity for reliable experimental performance.