Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentafluorophenyl)phenylphosphine, 97%

Catalog No. AAA1101401
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1101401 0.25 g
AAA1101403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1101401 Supplier Thermo Scientific Chemicals Supplier No. A1101401
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5074-71-5 | C18H5F10P | 442.20 g/mol

Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bis(pentafluorophenyl)phenylphosphine is used as a ligand involved in the palladium-catalyzed Heck reaction of iodobenzene and styrene in compressed carbon dioxide. It is also used in the polymerization of 1,3-butadiene with Co(2-ethyl hexanoate)2.

Solubility
Soluble in chloroform, dichloromethane, tetrahydrofuran, benzene, toluene, ethanol, methanol, and supercritical carbon dioxide.

Notes
Store in a cool place. Incompatible with strong oxidizing agents, strong reducing agents, strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5074-71-5
Molecular Formula C18H5F10P
Molecular Weight (g/mol) 442.20
MDL Number MFCD00000291
InChI Key OYNXPGGNQMSMTR-UHFFFAOYSA-N
Synonym bis pentafluorophenyl phenylphosphine, dftpp, decafluorotriphenylphosphine, bis perfluorophenyl phenyl phosphine, bis 2,3,4,5,6-pentafluorophenyl-phenylphosphane, phosphine,bis 2,3,4,5,6-pentafluorophenyl phenyl, bis 2,3,4,5,6-pentafluorophenyl phenyl phosphane, ultramark 443, bis pentafluorophenyl phenyl phosphite, bis pentafluorophenyl phenyl phosphine
PubChem CID 78757
IUPAC Name bis(2,3,4,5,6-pentafluorophenyl)-phenylphosphane
SMILES FC1=C(F)C(F)=C(P(C2=CC=CC=C2)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F

Specifications

Melting Point 59°C to 64°C
Boiling Point 105°C (0.3 mmHg)
Flash Point >110°C (230°F)
Quantity 0.25 g
Beilstein 6253897
Solubility Information Soluble in chloroform,dichloromethane,tetrahydrofuran,benzene,toluene,ethanol,methanol,and supercritical carbon dioxide.
Formula Weight 442.2
Percent Purity 97%
Chemical Name or Material Bis (pentafluorophenyl)phenylphosphine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 225-780-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.