Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate, 98%

Catalog No. AAH2776103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2776103 1 g
AAH2776106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2776103 Supplier Thermo Scientific Chemicals Supplier No. H2776103
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 79973-42-5 | C20H30CoF6P

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79973-42-5
Molecular Formula C20H30CoF6P
MDL Number MFCD00269818

Specifications

Melting Point ∽305°C (decomposition)
Quantity 1 g
UN Number UN3261
Formula Weight 474.36
Percent Purity 98%
Chemical Name or Material Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate
Hazard Category H314-H335-H350
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.