Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(pentamethylcyclopentadienyl)nickel

Catalog No. AA39238MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39238MD 250 mg
AA3923803 1 g
AA3923806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA39238MD Supplier Thermo Scientific Chemicals Supplier No. 039238MD
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 74507-63-4 | C20H30Ni | 329.153 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74507-63-4
Molecular Formula C20H30Ni
Molecular Weight (g/mol) 329.153
MDL Number MFCD00058704
InChI Key CHPLEWYRKUFKQP-UHFFFAOYSA-N
Synonym Decamethylnickelocene
PubChem CID 24943158
IUPAC Name nickel(2+);1,2,3,5,5-pentamethylcyclopenta-1,3-diene
SMILES CC1=[C-]C(C(=C1C)C)(C)C.CC1=[C-]C(C(=C1C)C)(C)C.[Ni+2]

Specifications

Packaging Ampule
Quantity 250 mg
Sensitivity Air and moisture sensitive
Formula Weight 329.17
Physical Form Crystalline
Chemical Name or Material Bis(pentamethylcyclopentadienyl)nickel
Hazard Category H350
Hazard Statement GHS H Statement
H350
May cause cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.