Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 25g, Bis[(pinacolato)boryl]methane, 97%, [78782-17-9], M13441 267.965 C13H26B2O4

Catalog No. 501455437
Encompass_Preferred

Compound: Bis[(pinacolato)boryl]methane

CAS#: 78782-17-9

MDL#: MFCD27977747

Background: A geminal bis(boronate) reagent used in organic synthesis to introduce two independently functionalizable boron centers on a single carbon. It is a versatile building block for iterative cross-coupling and other organoboron chemistry.

Storage: Store at room temperature, under dry conditions.

Catalog No. 50-145-5437 Supplier Frontier Specialty Chemicals Supplier No. M1344125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Bis[(pinacolato)boryl]methane

CAS#: 78782-17-9

MDL#: MFCD27977747

Background: A geminal bis(boronate) reagent used in organic synthesis to introduce two independently functionalizable boron centers on a single carbon. It is a versatile building block for iterative cross-coupling and other organoboron chemistry.

Storage: Store at room temperature, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.