Learn More
Strem, An Ascensus Company CAS# 73183-34-3. 2g. Bis(pinacolato)diboron, 99%. MFCD00799570
SDP
Catalog No. 501385323
Encompass
CAS# 73183-34-3. 2g. Bis(pinacolato)diboron, 99%. MFCD00799570. Molecular Weight: 253.94. Molecular Formula: C12H24B2O4. Color/form: white to off-white xtl. Strem# 05-0030. http://www.strem.com/catalog/v/05-0030/
Catalog No. 50-138-5323 Supplier Strem, An Ascensus Company Supplier No. 0500302G
CAS# 73183-34-3. 2g. Bis(pinacolato)diboron, 99%. MFCD00799570. Molecular Weight: 253.94. Molecular Formula: C12H24B2O4. Color/form: white to off-white xtl. Strem# 05-0030. http://www.strem.com/catalog/v/05-0030/