Learn More
Medchemexpress LLC Bis(pinacolato)diborane | 73183-34-3 | 99.9% | C12H24B2O4 | 25 G
SDP
Catalog No. 5000358902
Encompass_Preferred
Bis(pinacolato)diborane is an organoboron reagent that serves as a source of the [Bpin] moiety with pronounced nucleophilic character. It is utilized in the synthesis of various organoboranes, including organoboron compounds and arylboronates.
  • Acts as a source of the [Bpin] moiety with nucleophilic character
  • Used in the synthesis of organoboron compounds and arylboronates
  • Reacts with p-benzyne to produce tetraborylated products
Catalog No. 50-003-58902 Supplier Medchemexpress LLC Supplier No. 25GHY79444
Bis(pinacolato)diborane is an organoboron reagent that serves as a source of the [Bpin] moiety with pronounced nucleophilic character. It is utilized in the synthesis of various organoboranes, including organoboron compounds and arylboronates.
  • Acts as a source of the [Bpin] moiety with nucleophilic character
  • Used in the synthesis of organoboron compounds and arylboronates
  • Reacts with p-benzyne to produce tetraborylated products