Learn More
Medchemexpress LLC Bis(pinacolato)diborane | 73183-34-3 | 99.97% | C12H24B2O4 | 10 G
SDP
Supplier:  Medchemexpress LLC HY7944410G
Encompass_Preferred
Bis(pinacolato)diborane is an organoboron reagent that serves as a source of the [Bpin] moiety with pronounced nucleophilic character. It is used in the synthesis of various organoboranes, including organoboron compounds and arylboronates. This white to off-white solid reacts with p-benzyne to produce tetraborylated products and can tetraborate p-benzoyne generated from 1,2-diethynylbenzene derivatives for in vitro applications. It is recommended for research use only.
  • Serves as a source of the [Bpin] moiety with pronounced nucleophilic character.
  • Used in the synthesis of various organoboranes, including organoboron compounds and arylboronates.
  • Reacts with p-benzyne to produce tetraborylated products.
  • Can tetraborate p-benzoyne generated from 1,2-diethynylbenzene derivatives for in vitro applications.
Catalog No. 50-004-32718