Learn More
Medchemexpress LLC Bis(pinacolato)diborane | 73183-34-3 | 99.97% | C12H24B2O4 | 100 G
SDP
Catalog No. 5000433120
Encompass_Preferred
Bis(pinacolato)diborane is an organoboron reagent. It functions as a source of the [Bpin] moiety, which exhibits a pronounced nucleophilic character. This makes it valuable in the synthesis of various organoboranes, including organoboron compounds and arylboronates. Additionally, Bis(pinacolato)diborane is known to react with p-benzyne, leading to the production of tetraborylated products through a series of steps involving addition and 1,2-migration of the boryl group.
  • Organoboron reagent
  • Source of nucleophilic [Bpin] moiety
  • Used in the synthesis of various organoboranes, including organoboron compounds and arylboronates
  • Reacts with p-benzyne to produce tetraborylated products
Catalog No. 50-004-33120 Supplier Medchemexpress LLC Supplier No. HY79444100G
Bis(pinacolato)diborane is an organoboron reagent. It functions as a source of the [Bpin] moiety, which exhibits a pronounced nucleophilic character. This makes it valuable in the synthesis of various organoboranes, including organoboron compounds and arylboronates. Additionally, Bis(pinacolato)diborane is known to react with p-benzyne, leading to the production of tetraborylated products through a series of steps involving addition and 1,2-migration of the boryl group.
  • Organoboron reagent
  • Source of nucleophilic [Bpin] moiety
  • Used in the synthesis of various organoboranes, including organoboron compounds and arylboronates
  • Reacts with p-benzyne to produce tetraborylated products