Learn More
Medchemexpress LLC Bis(pinacolato)diborane | 73183-34-3 | 99.97% | C12H24B2O4 | 5 G
SDP
Catalog No. 5000433250
Encompass_Preferred
Bis(pinacolato)diborane is an organoboron reagent that serves as a source of the [Bpin] moiety with pronounced nucleophilic character. It is utilized in the synthesis of various organoboranes, including organoboron compounds and arylboronates. This compound reacts with p-benzyne to produce tetraborylated products through a series of steps involving addition and 1,2-migration of the boryl group.
  • Source of [Bpin] moiety with nucleophilic character
  • Used in the synthesis of various organoboranes
  • Reacts with p-benzyne to produce tetraborylated products
  • Suitable for in vitro biological activity studies
Catalog No. 50-004-33250 Supplier Medchemexpress LLC Supplier No. HY794445G
Bis(pinacolato)diborane is an organoboron reagent that serves as a source of the [Bpin] moiety with pronounced nucleophilic character. It is utilized in the synthesis of various organoboranes, including organoboron compounds and arylboronates. This compound reacts with p-benzyne to produce tetraborylated products through a series of steps involving addition and 1,2-migration of the boryl group.
  • Source of [Bpin] moiety with nucleophilic character
  • Used in the synthesis of various organoboranes
  • Reacts with p-benzyne to produce tetraborylated products
  • Suitable for in vitro biological activity studies