Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Matrix Scientific Bis(pinacolato)diboron, 73183-34-3, MFCD00799570, 25g

Catalog No. 500117300
Encompass_Preferred

Molecular Formula C12H24B2O4, Purity 98%, Molecular Weight 253.94, Melting Point 115-117°

Catalog No. 50-011-7300 Supplier Matrix Scientific Supplier No. 00488925G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Formula C12H24B2O4, Purity 98%, Molecular Weight 253.94, Melting Point 115-117°

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.