Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Frontier Specialty Chemicals 1kg, Bis(pinacolato)diboron, 99%, [73183-34-3], D6878 253.94 C12H24B2O4

Catalog No. 501409093
Encompass_Preferred

Compound: Bis(pinacolato)diboron

CAS#: 73183-34-3

MDL#: MFCD00799570

Background: A versatile reagent and specialty chemical used, with iridium or other transition metal catalysts, for the C-H borylation of arenes, heteroarenes, and benzylic positions of alkylarenes to furnish aryl-, heteroaryl-, and benzylboron compounds. It also reacts under metal-catalyzed conditions with various functional groups and scaffolds, including polymers and terminal alkenes to generate diols.

Storage: Store at -20°C, under dry conditions.

Catalog No. 50-140-9093 Supplier Frontier Specialty Chemicals Supplier No. D68781KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Compound: Bis(pinacolato)diboron

CAS#: 73183-34-3

MDL#: MFCD00799570

Background: A versatile reagent and specialty chemical used, with iridium or other transition metal catalysts, for the C-H borylation of arenes, heteroarenes, and benzylic positions of alkylarenes to furnish aryl-, heteroaryl-, and benzylboron compounds. It also reacts under metal-catalyzed conditions with various functional groups and scaffolds, including polymers and terminal alkenes to generate diols.

Storage: Store at -20°C, under dry conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.