Learn More
Medchemexpress LLC Bis-propargyl-PEG4 | 159428-42-9 | 95.6% | 5 G
SDP
Catalog No. 5000231836
Encompass_Preferred
Bis-propargyl-PEG4 is a PEG-based bifunctional linker featuring terminal propargyl (alkyne) groups, designed for conjugation chemistry and PROTAC construction. It provides a short, flexible spacer for joining two ligands via click chemistry.
  • Bifunctional PEG4 linker with terminal alkyne groups.
  • Enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Useful for PROTAC synthesis and dimer formation.
  • Low molecular weight (270.32 g/mol) for compact linker design.
  • Stable under recommended storage conditions when kept dry and refrigerated.
Catalog No. 50-002-31836 Supplier Medchemexpress LLC Supplier No. HY1203975G
Bis-propargyl-PEG4 is a PEG-based bifunctional linker featuring terminal propargyl (alkyne) groups, designed for conjugation chemistry and PROTAC construction. It provides a short, flexible spacer for joining two ligands via click chemistry.
  • Bifunctional PEG4 linker with terminal alkyne groups.
  • Enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Useful for PROTAC synthesis and dimer formation.
  • Low molecular weight (270.32 g/mol) for compact linker design.
  • Stable under recommended storage conditions when kept dry and refrigerated.