Learn More
Medchemexpress LLC Bis-propargyl-PEG8 100mg | 1092554-87-4 | 446.53 | C22H38O9 | 100 MG
SDP
Catalog No. 5000228743
Encompass_Preferred
Bis-propargyl-PEG8 is a PEG-based linker bearing two terminal propargyl (alkyne) groups designed for click-chemistry conjugation. It provides a flexible PEG8 spacer for use in PROTAC synthesis and other bioconjugation workflows, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) between azide- and alkyne-functionalized partners.
  • Contains two terminal alkyne groups for CuAAC click chemistry.
  • Peg8 spacer improves solubility and provides flexible linker length.
  • Suitable for synthesis of PROTACs and other bioconjugates.
  • Available in laboratory-scale pack sizes for research use.
  • Provided as a ready-to-use reagent for organic synthesis workflows.
Catalog No. 50-002-28743 Supplier Medchemexpress LLC Supplier No. HY115414100MG
Bis-propargyl-PEG8 is a PEG-based linker bearing two terminal propargyl (alkyne) groups designed for click-chemistry conjugation. It provides a flexible PEG8 spacer for use in PROTAC synthesis and other bioconjugation workflows, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAC) between azide- and alkyne-functionalized partners.
  • Contains two terminal alkyne groups for CuAAC click chemistry.
  • Peg8 spacer improves solubility and provides flexible linker length.
  • Suitable for synthesis of PROTACs and other bioconjugates.
  • Available in laboratory-scale pack sizes for research use.
  • Provided as a ready-to-use reagent for organic synthesis workflows.