Learn More
Medchemexpress LLC Bis-propargyl-PEG8 10mg | 1092554-87-4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11541410MG
Encompass_Preferred
Bis-propargyl-PEG8 is a PEG8-based bifunctional linker bearing terminal propargyl (alkyne) groups, used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAC) in PROTAC and bioconjugation synthesis.
  • Facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Suitable for PROTAC linker construction and bioconjugation chemistry.
  • PEG8 spacer improves solubility and provides flexible linkage length.
  • Contains two terminal alkynes for dual conjugation sites.
  • Supplied in small, pre-weighed mg quantities for research applications.
Catalog No. 50-002-28455