Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-propargyl-PEG8 250mg | 1092554-87-4 | 250 MG
SDP

Supplier:  Medchemexpress LLC HY115414250MG

Encompass_Preferred

Bis-propargyl-PEG8 is a PEG-based PROTAC linker with two terminal propargyl (alkyne) groups used as a click-chemistry reagent to join azide-bearing fragments via copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is supplied in small-scale laboratory pack sizes for research use.

  • Contains two terminal alkyne (propargyl) groups.
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • PEG8 spacer provides a flexible, hydrophilic linker.
  • Supplied in multiple small laboratory pack sizes.
  • Molecular formula C22H38O9; molecular weight 446.53.

Catalog No. 50-002-28173


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.