Learn More
Medchemexpress LLC Bis-propargyl-PEG8 50mg | 1092554-87-4 | 50 MG
SDP
Catalog No. 5000307657
Encompass_Preferred
Bis-propargyl-PEG8 is a polyethylene glycol-based bifunctional linker bearing terminal propargyl (alkyne) groups. It functions as a flexible PEG8 spacer for click-chemistry conjugations, PROTAC construction, and other bioconjugation applications.
  • Contains two terminal propargyl (alkyne) groups for CuAAC click reactions.
  • PEG8 spacer provides flexibility and increased hydrophilicity.
  • Applicable to PROTAC and ADC linker synthesis and general bioconjugation workflows.
  • Supplied as a solid, commonly offered in milligram quantities such as 50 mg.
  • Characterized by chemical formula C22H38O9 and molecular weight 446.53 g·mol-1.
Catalog No. 50-003-07657 Supplier Medchemexpress LLC Supplier No. HY11541450MG
Bis-propargyl-PEG8 is a polyethylene glycol-based bifunctional linker bearing terminal propargyl (alkyne) groups. It functions as a flexible PEG8 spacer for click-chemistry conjugations, PROTAC construction, and other bioconjugation applications.
  • Contains two terminal propargyl (alkyne) groups for CuAAC click reactions.
  • PEG8 spacer provides flexibility and increased hydrophilicity.
  • Applicable to PROTAC and ADC linker synthesis and general bioconjugation workflows.
  • Supplied as a solid, commonly offered in milligram quantities such as 50 mg.
  • Characterized by chemical formula C22H38O9 and molecular weight 446.53 g·mol-1.