Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-propargyl-PEG8 5mg | 1092554-87-4 | 5 MG
SDP

Catalog No. 5000228172
Encompass_Preferred

Bis-propargyl-PEG8 is a PEG-based bifunctional linker with two terminal propargyl (alkyne) groups used as a click-chemistry reagent for bioconjugation and PROTAC synthesis.

  • Contains two terminal propargyl (alkyne) groups for CuAAC reactions.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • PEG8 spacer provides hydrophilicity and flexible linkage.
  • Molecular formula C22H38O9 and molecular weight 446.53 g·mol⁻¹.
  • Available in small research quantities such as 5 MG.

Catalog No. 50-002-28172 Supplier Medchemexpress LLC Supplier No. HY1154145MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-propargyl-PEG8 is a PEG-based bifunctional linker with two terminal propargyl (alkyne) groups used as a click-chemistry reagent for bioconjugation and PROTAC synthesis.

  • Contains two terminal propargyl (alkyne) groups for CuAAC reactions.
  • Suitable for copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • PEG8 spacer provides hydrophilicity and flexible linkage.
  • Molecular formula C22H38O9 and molecular weight 446.53 g·mol⁻¹.
  • Available in small research quantities such as 5 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.