Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis[rhodium(alpha,alpha,alpha',alpha'-tetramethyl-1,3-benzenedipropionic Acid)] 96.0+%, TCI America™
SDP

Catalog No. B4549100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4549100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4549100MG Supplier TCI America Supplier No. B4549100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 819050-89-0
Molecular Formula C32H40O8Rh2
Molecular Weight (g/mol) 758.48
MDL Number MFCD08457636
InChI Key AHCPGXZDVSBJII-UHFFFAOYSA-J
Synonym Rh2(esp)2
PubChem CID 131885550
IUPAC Name bis(λ²-rhodium(2+)) bis(3-[3-(2-carboxylato-2,2-dimethylethyl)phenyl]-2,2-dimethylpropanoate)
SMILES [Rh++].[Rh++].CC(C)(CC1=CC(CC(C)(C)C([O-])=O)=CC=C1)C([O-])=O.CC(C)(CC1=CC(CC(C)(C)C([O-])=O)=CC=C1)C([O-])=O

Specifications

Color Green-Yellow
Quantity 100 mg
Formula Weight 758.48
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Bis[rhodium(alpha,alpha,alpha′,alpha′-tetramethyl-1,3-benzenedipropionic Acid)]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.