Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (-)-Bis[(S)-1-Phenylethyl]Amine

Catalog No. 43381010G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-381-010G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 43-381-010G Supplier Sigma Aldrich Supplier No. 43381010G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(-)-Bis[(S)-1-phenylethyl]amine can be used as:A key intermediate in the synthesis of phosphoramidite (Feringa) ligands.A reactant in the preparation of C(18)-C(24) unit of a novel macrolide compound lasonolide A.A starting material in the synthesis of tacrine-huperzine A hybrid.

Specifications

CAS 76769-55-6
Melting Point 127°C to 130°C
Molecular Formula C14H22O8
Linear Formula [(CH3)3CCO2CH(CO2H)-]2
Refractive Index n20/D 1.5525
MDL Number MFCD00015634
Quantity 5 g
Synonym (-)-Bis[(S)-alpha-methylbenzyl]amine; [S-(R,R)]-(-)-Bis(alpha-methylbenzyl)amine
Molecular Weight (g/mol) 318.32 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.