Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-SS-C3-NHS ester | 98604-88-7 | 98.0% | 432.47 | C16H20N2O8S2 | 1 G
SDP

Catalog No. 5000202407
Encompass_Preferred

Bis-SS-C3-NHS ester is a disulfide-cleavable N-hydroxysuccinimide (NHS) ester linker used for bioconjugation and antibody-drug conjugate (ADC) synthesis. It couples to primary amines via NHS-ester chemistry and releases payloads under reducing intracellular conditions.

  • Molecular weight: 432.47
  • Chemical formula: C16H20N2O8S2
  • Cas number: 98604-88-7
  • Purity: 98.0%
  • Appearance: white to off-white solid
  • Reactive group: NHS ester for coupling to primary amines
  • Application: cleavable disulfide linker for ADC synthesis and bioconjugation
  • Available pack size: 1 g

Catalog No. 50-002-02407 Supplier Medchemexpress LLC Supplier No. HY1335841G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-SS-C3-NHS ester is a disulfide-cleavable N-hydroxysuccinimide (NHS) ester linker used for bioconjugation and antibody-drug conjugate (ADC) synthesis. It couples to primary amines via NHS-ester chemistry and releases payloads under reducing intracellular conditions.

  • Molecular weight: 432.47
  • Chemical formula: C16H20N2O8S2
  • Cas number: 98604-88-7
  • Purity: 98.0%
  • Appearance: white to off-white solid
  • Reactive group: NHS ester for coupling to primary amines
  • Application: cleavable disulfide linker for ADC synthesis and bioconjugation
  • Available pack size: 1 g

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.