Learn More
Medchemexpress LLC Bis-SS-C3-NHS ester | 98604-88-7 | 98.0% | 432.47 g/mol | C16H20N2O8S2 | 10 MG
SDP
Catalog No. 5000203696
Encompass_Preferred
Bis-SS-C3-NHS ester is a cleavable disulfide-containing N-hydroxysuccinimide (NHS) ester linker used for bioconjugation. It reacts with primary amines to form stable amide bonds, and the internal disulfide provides a reducible linkage for intracellular payload release, making it suitable for antibody-drug conjugate synthesis and conjugation method development.
  • Cleavable disulfide linker for controlled payload release
  • N-hydroxysuccinimide (NHS) ester reactive toward primary amines
  • High purity suitable for research-scale conjugation
  • Available in multiple small-scale pack sizes for method development
  • Stable when stored sealed at -20°C and protected from moisture
Catalog No. 50-002-03696 Supplier Medchemexpress LLC Supplier No. HY13358410MG
Bis-SS-C3-NHS ester is a cleavable disulfide-containing N-hydroxysuccinimide (NHS) ester linker used for bioconjugation. It reacts with primary amines to form stable amide bonds, and the internal disulfide provides a reducible linkage for intracellular payload release, making it suitable for antibody-drug conjugate synthesis and conjugation method development.
  • Cleavable disulfide linker for controlled payload release
  • N-hydroxysuccinimide (NHS) ester reactive toward primary amines
  • High purity suitable for research-scale conjugation
  • Available in multiple small-scale pack sizes for method development
  • Stable when stored sealed at -20°C and protected from moisture