Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-SS-C3-NHS ester | 98604-88-7 | 98.0% | 432.47 g/mol | C16H20N2O8S2 | 100 MG
SDP

Catalog No. 5000202406
Encompass_Preferred

Bis-SS-C3-NHS ester is a cleavable antibody-drug conjugate (ADC) linker containing a disulfide-based spacer and N-hydroxysuccinimide (NHS) ester reactive groups for amine coupling. It is used to attach cytotoxic payloads to antibodies and enable controlled payload release inside target cells.

  • Molecular formula: C16H20N2O8S2.
  • Molecular weight: 432.47 g/mol.
  • Cas number: 98604-88-7.
  • Purity: 98.0%.
  • Appearance: white to off-white solid.
  • Storage: in solvent, -80°C for up to 6 months; -20°C for up to 1 month; store sealed and away from moisture.
  • Available in multiple sizes, including 100 mg, for research use.

Catalog No. 50-002-02406 Supplier Medchemexpress LLC Supplier No. HY133584100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-SS-C3-NHS ester is a cleavable antibody-drug conjugate (ADC) linker containing a disulfide-based spacer and N-hydroxysuccinimide (NHS) ester reactive groups for amine coupling. It is used to attach cytotoxic payloads to antibodies and enable controlled payload release inside target cells.

  • Molecular formula: C16H20N2O8S2.
  • Molecular weight: 432.47 g/mol.
  • Cas number: 98604-88-7.
  • Purity: 98.0%.
  • Appearance: white to off-white solid.
  • Storage: in solvent, -80°C for up to 6 months; -20°C for up to 1 month; store sealed and away from moisture.
  • Available in multiple sizes, including 100 mg, for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.