Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bis-sulfone-PEG3-azide | 1802908-01-5 | MFCD29052187 | 97.8% | 700.82 g/mol | C33H40N4O9S2 | 5 MG
SDP

Catalog No. 5000203489
Encompass_Preferred

Bis-sulfone-PEG3-azide is a PEG-based, cleavable linker bearing an azide functional group used for bioconjugation and ADC/PROTAC linker synthesis. It enables click chemistry conjugation (CuAAC and SPAAC) between azide and alkyne functional groups, and is supplied as a small-molecule reagent for research use.

  • Cleavable ADC and PROTAC linker for bioconjugation.
  • Contains an azide group for CuAAC and strain-promoted cycloaddition (SPAAC) reactions.
  • Molecular weight approximately 700.82 g/mol; molecular formula C33H40N4O9S2.
  • CAS number 1802908-01-5 for unambiguous chemical identification.
  • Available in a 5 mg pack; larger sizes available by quote.
  • Storage: dry, -20°C (stable ~3 years); in solvent, -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-03489 Supplier Medchemexpress LLC Supplier No. HY1387455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bis-sulfone-PEG3-azide is a PEG-based, cleavable linker bearing an azide functional group used for bioconjugation and ADC/PROTAC linker synthesis. It enables click chemistry conjugation (CuAAC and SPAAC) between azide and alkyne functional groups, and is supplied as a small-molecule reagent for research use.

  • Cleavable ADC and PROTAC linker for bioconjugation.
  • Contains an azide group for CuAAC and strain-promoted cycloaddition (SPAAC) reactions.
  • Molecular weight approximately 700.82 g/mol; molecular formula C33H40N4O9S2.
  • CAS number 1802908-01-5 for unambiguous chemical identification.
  • Available in a 5 mg pack; larger sizes available by quote.
  • Storage: dry, -20°C (stable ~3 years); in solvent, -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.