Learn More
Medchemexpress LLC Bis(sulfosuccinimidyl) suberate | 82436-77-9 | 99.8% | 528.47 g/mol | C16H20N2O14S2 | 5 MG
SDP
Catalog No. 5000202776
Encompass_Preferred
BS3 crosslinker (bis(sulfosuccinimidyl) suberate) is a water-soluble, non-cleavable, homobifunctional NHS-ester reagent used to covalently link primary amines. It is commonly applied for protein-protein conjugation, antibody conjugation workflows, and general bioconjugation where stable amine-to-amine linkages are required. The reagent is provided with documented purity and handling instructions to support reproducible results.
  • Water-soluble homobifunctional NHS-ester for amine-to-amine crosslinking.
  • Non-cleavable linker suitable for producing stable conjugates.
  • Reacts with lysine residues under physiological conditions for mild conjugation.
  • High purity supports reproducible bioconjugation performance.
  • Available in multiple pack sizes for flexible scaling of experiments.
  • Storage and handling guidance provided to preserve reagent stability.
Catalog No. 50-002-02776 Supplier Medchemexpress LLC Supplier No. HY1243295MG
BS3 crosslinker (bis(sulfosuccinimidyl) suberate) is a water-soluble, non-cleavable, homobifunctional NHS-ester reagent used to covalently link primary amines. It is commonly applied for protein-protein conjugation, antibody conjugation workflows, and general bioconjugation where stable amine-to-amine linkages are required. The reagent is provided with documented purity and handling instructions to support reproducible results.
  • Water-soluble homobifunctional NHS-ester for amine-to-amine crosslinking.
  • Non-cleavable linker suitable for producing stable conjugates.
  • Reacts with lysine residues under physiological conditions for mild conjugation.
  • High purity supports reproducible bioconjugation performance.
  • Available in multiple pack sizes for flexible scaling of experiments.
  • Storage and handling guidance provided to preserve reagent stability.