Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(sulfur Dioxide)-1,4-diazabicyclo[2.2.2]octane Adduct 97.0+%, TCI America™
SDP

Catalog No. B39601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B39601G 1 g
B39605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B39601G Supplier TCI America Supplier No. B39601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 119752-83-9
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
InChI Key RWISEVUOFYXWFO-UHFFFAOYSA-N
Synonym 1,4-diazabicyclo 2.2.2 octane-1,4-diium-1,4-disulfinate, 1,4-diazoniabicyclo 2.2.2 octane-1,4-disulfinate, 1,4-diazabicyclo 2.2.2 octane-1,4-diium-1,4-disulf, 1,4-diazabicyclo 2.2.2 octane bis sulfur dioxide adduct, 1,4-diazoniabicyclo 2.2.2 octane-1,4-diylbis dioxosulfur vi, 1,4-diazabicyclo 2.2.2 octane bis sulfur dioxide adduct sulfur, elemental analysis
PubChem CID 75176251
IUPAC Name 1,4-diazoniabicyclo[2.2.2]octane-1,4-disulfinate
SMILES C1C[N+]2(CC[N+]1(CC2)S(=O)[O-])S(=O)[O-]

Specifications

Melting Point 180°C
Color White
Quantity 1 g
Formula Weight 240.29
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Bis(sulfur Dioxide)-1,4-diazabicyclo[2.2.2]octane Adduct
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.